Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079977.2 | NP_001073446.2 | 10 - 11 | 1394 | 131 | |
XM_005165515.4 | XP_005165572.1 | 10 - 11 | 1369 | 131 | ||
GenBank mRNA | BC117622.1 | - | 10 - 11 | 1398 | 131 | |
BC155135.1 | - | 10 - 11 | 1369 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |