Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atpaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082821.1 | NP_001076290.1 | 2 - 3 | 429 | 142 | |
GenBank mRNA | BC134167.1 | - | 2 - 3 | 429 | 142 | |
BC153513.1 | - | 2 - 3 | 439 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |