Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083002.1 | NP_001076471.1 | 2 - 3 | 770 | 66 | |
XM_005163128.4 | XP_005163185.1 | 2 - 3 | 962 | 66 | ||
XM_009298079.3 | XP_009296354.1 | 2 - 3 | 753 | 66 | ||
GenBank mRNA | BC131867.1 | - | 2 - 3 | 770 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |