Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfsd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083056.1 | NP_001076525.1 | 2 - 3 | 1560 | 63 | |
XM_005161679.4 | XP_005161736.1 | 3 - 4 | 2529 | 63 | ||
XM_021469281.1 | XP_021324956.1 | 2 - 3 | 1560 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |