Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfsd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083056.1 | NP_001076525.1 | 6 - 7 | 2208 | 79 | |
XM_005161679.4 | XP_005161736.1 | 7 - 8 | 3177 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |