Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113621.1 | NP_001107093.1 | 2 - 3 | 476 | 86 | |
GenBank mRNA | BC154444.1 | - | 2 - 3 | 476 | 86 | |
BC171521.1 | - | 2 - 3 | 471 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |