Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp4t8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199216.2 | NP_954686.2 | 8 - 9 | 1126 | 90 | |
GenBank mRNA | BC056734.1 | - | 8 - 9 | 1094 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |