Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200088.1 | NP_956382.1 | 6 - 7 | 763 | 144 | |
GenBank mRNA | AY648836.1 | - | 6 - 7 | 744 | 144 | |
BC053230.1 | - | 6 - 7 | 763 | 144 | ||
BC154333.1 | - | 5 - 6 | 784 | 144 | ||
BC171713.1 | - | 6 - 7 | 748 | 144 | ||
BC171715.1 | - | 6 - 7 | 748 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |