Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rundc3ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200149.1 | NP_956443.1 | 7 - 8 | 809 | 81 | |
GenBank mRNA | BC044502.1 | - | 7 - 8 | 809 | 81 | |
BC044540.1 | - | 7 - 8 | 809 | 81 | ||
BC155785.1 | - | 6 - 7 | 992 | 81 | ||
BC165804.1 | - | 6 - 7 | 638 | 81 | ||
EF646262.1 | - | 6 - 7 | 618 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |