Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tuba8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200691.1 | NP_956985.1 | 2 - 3 | 419 | 113 | |
GenBank mRNA | AY391473.1 | - | 2 - 3 | 293 | 113 | |
BC059428.1 | - | 2 - 3 | 419 | 113 | ||
BC062826.1 | - | 2 - 3 | 366 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |