Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122971.1 | NP_001116443.1 | 6 - 7 | 936 | 90 | |
XM_009297890.2 | XP_009296165.1 | 6 - 7 | 1159 | 90 | ||
XM_021470456.1 | XP_021326131.1 | 4 - 5 | 663 | 90 | ||
GenBank mRNA | AJ544829.2 | - | 6 - 7 | 936 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |