Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: th | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131149.1 | NP_571224.1 | 11 - 12 | 1177 | 120 | |
GenBank mRNA | BC163630.1 | - | 11 - 12 | 1244 | 120 | |
BC163639.1 | - | 11 - 12 | 1244 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |