Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201319.2 | NP_958476.2 | 9 - 10 | 1797 | 85 | |
GenBank mRNA | BC055651.1 | - | 10 - 11 | 1206 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |