Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212738.1 | NP_997903.1 | 3 - 4 | 277 | 77 | |
GenBank mRNA | AY289606.1 | - | 3 - 4 | 277 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |