Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081893.1 | NP_001075362.1 | 3 - 4 | 534 | 92 | |
GenBank mRNA | AJ428063.1 | - | 1 - 2 | 304 | 92 | |
AY290704.1 | - | 3 - 4 | 534 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |