Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099764.1 | NP_001093234.1 | 2 - 3 | 75 | 112 | |
GenBank mRNA | AB163433.1 | - | 2 - 3 | 75 | 112 | |
AY587560.1 | - | 2 - 3 | 64 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |