Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111299.2 | NP_001104769.2 | 3 - 4 | 1087 | 72 | |
GenBank mRNA | AF368321.1 | - | 1 - 2 | 94 | 72 | |
EU025848.1 | - | 3 - 4 | 1087 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Equus caballus Probe spans exons |