Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111342.1 | NP_001104812.1 | 3 - 4 | 367 | 120 | |
GenBank mRNA | EU184658.1 | - | 3 - 4 | 367 | 120 | |
HQ596524.1 | - | 3 - 4 | 577 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |