Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114942.1 | NP_001108414.1 | 3 - 4 | 515 | 86 | |
GenBank mRNA | EF682146.1 | - | 3 - 4 | 515 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |