Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163968.2 | NP_001157440.2 | 2 - 3 | 490 | 86 | |
GenBank mRNA | DQ409212.1 | - | 1 - 2 | 109 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |