Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143791.1 | NP_001137263.1 | 2 - 3 | 190 | 73 | |
XM_023616897.1 | XP_023472665.1 | 1459 | 73 | |||
GenBank mRNA | DQ889711.1 | - | 2 - 3 | 182 | 73 | |
EF645663.1 | - | 2 - 3 | 190 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Equus caballus Probe spans exons |