Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085433.2 | NP_001078902.1 | 2 - 3 | 236 | 145 | |
GenBank mRNA | EU438768.1 | - | 2 - 3 | 234 | 145 | |
JQ432544.1 | - | 2 - 3 | 205 | 145 | ||
L06009.1 | - | 2 - 3 | 205 | 145 | ||
X69393.1 | - | 236 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |