Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205192.1 | NP_001192121.1 | 2 - 3 | 575 | 77 | |
XM_023644173.1 | XP_023499941.1 | 1116 | 77 | |||
XM_023644175.1 | XP_023499943.1 | 626 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |