Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205261.1 | NP_001192190.1 | 4 - 5 | 1349 | 74 | |
XM_014730313.2 | XP_014585799.2 | 1338 | 74 | |||
XM_014730314.2 | XP_014585800.2 | 1314 | 74 | |||
XM_014730317.2 | XP_014585803.2 | 1188 | 74 | |||
GenBank mRNA | CX593658.1 | - | 3 - 4 | 470 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |