Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 10 - 11 | 452 | 87 | |
XM_014738515.2 | XP_014594001.1 | 463 | 87 | |||
GenBank mRNA | GU196267.1 | - | 10 - 11 | 452 | 87 | |
KP658381.1 | - | 11 - 12 | 459 | 87 | ||
KP658382.1 | - | 9 - 10 | 408 | 87 | ||
KT368778.1 | - | 11 - 12 | 453 | 87 | ||
KT368779.1 | - | 9 - 10 | 402 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |