Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198620.1 | NP_001185549.1 | 4 - 5 | 308 | 76 | |
GenBank mRNA | BX929583.1 | - | 3 - 4 | 209 | 76 | |
BX931100.2 | - | 4 - 5 | 336 | 76 | ||
BX932671.2 | - | 4 - 5 | 292 | 76 | ||
BX933955.1 | - | 4 - 5 | 308 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |