Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277999.1 | NP_001264928.1 | 1 - 2 | 89 | 90 | |
XM_015285101.2 | XP_015140587.1 | 2 - 3 | 119 | 90 | ||
XM_015285102.2 | XP_015140588.1 | 2 - 3 | 97 | 90 | ||
GenBank mRNA | BX930018.1 | - | 1 - 2 | 101 | 90 | |
BX934024.1 | - | 1 - 2 | 89 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |