Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277776.1 | NP_001264705.1 | 2 - 3 | 369 | 68 | |
NM_001277777.1 | NP_001264706.1 | 2 - 3 | 98 | 68 | ||
GenBank mRNA | BU363480.1 | - | 2 - 3 | 243 | 68 | |
BX929634.2 | - | 2 - 3 | 243 | 68 | ||
BX933235.1 | - | 2 - 3 | 226 | 68 | ||
BX935210.2 | - | 2 - 3 | 369 | 68 | ||
CN234998.1 | - | 2 - 3 | 101 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |