Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX929924.2 | - | 1 | 1147 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |