Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424952.5 | XP_424952.1 | 8 - 9 | 1184 | 69 | |
GenBank mRNA | CR385989.1 | - | 6 - 7 | 691 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |