Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039263.2 | NP_001034352.2 | 1 - 2 | 64 | 102 | |
XM_015294144.2 | XP_015149630.1 | 1 - 2 | 57 | 102 | ||
GenBank mRNA | AJ456913.1 | - | 1 - 2 | 65 | 102 | |
CR387393.1 | - | 1 - 2 | 51 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |