Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX934603.1 | - | 5 | 588 | 111 | |
BX936164.2 | - | 1 | 253 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |