Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277110.1 | NP_001264039.1 | 6 - 7 | 844 | 77 | |
NM_001277111.1 | NP_001264040.1 | 6 - 7 | 844 | 77 | ||
GenBank mRNA | BX929789.2 | - | 5 - 6 | 692 | 77 | |
CR390312.1 | - | 2 - 3 | 191 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |