Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271930.3 | NP_001258859.1 | 7 - 8 | 592 | 86 | |
NR_073522.3 | 6 - 7 | 563 | 86 | |||
GenBank mRNA | BX932147.1 | - | 2 - 3 | 577 | 86 | |
BX936070.2 | - | 7 - 8 | 592 | 86 | ||
CR405775.1 | - | 6 - 7 | 572 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |