Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277169.1 | NP_001264098.1 | 1 - 2 | 156 | 101 | |
GenBank mRNA | BX930961.1 | - | 1 - 2 | 136 | 101 | |
BX931234.1 | - | 1 - 2 | 125 | 101 | ||
BX932485.1 | - | 1 - 2 | 116 | 101 | ||
CR733399.1 | - | 1 - 2 | 92 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |