Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277465.1 | NP_001264394.1 | 5 | 781 | 74 | |
NM_001277467.1 | NP_001264396.1 | 4 | 697 | 74 | ||
NM_001277468.1 | NP_001264397.1 | 3 | 603 | 74 | ||
GenBank mRNA | BX929949.2 | - | 4 | 692 | 74 | |
BX931358.2 | - | 5 | 773 | 74 | ||
BX933179.2 | - | 2 | 74 | |||
BX933533.2 | - | 5 | 663 | 74 | ||
BX933747.1 | - | 1 | 350 | 74 | ||
CR353858.1 | - | 3 | 594 | 74 | ||
CR386803.1 | - | 5 | 759 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |