Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164350.1 | NP_001157822.1 | 1 - 2 | 143 | 86 | |
GenBank mRNA | BX950784.2 | - | 1 - 2 | 145 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |