Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015290549.2 | XP_015146035.1 | 2 - 3 | 135 | 69 | |
XM_422290.4 | XP_422290.2 | 2 - 3 | 506 | 69 | ||
GenBank mRNA | CR385470.1 | - | 2 - 3 | 207 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |