Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277175.1 | NP_001264104.1 | 1 - 2 | 176 | 81 | |
GenBank mRNA | BX929687.1 | - | 1 - 2 | 53 | 81 | |
BX933093.2 | - | 1 - 2 | 106 | 81 | ||
BX934630.1 | - | 1 - 2 | 141 | 81 | ||
BX935510.1 | - | 1 - 2 | 99 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |