Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278089.1 | NP_001265018.1 | 5 - 6 | 838 | 139 | |
GenBank mRNA | BX950347.1 | - | 5 - 6 | 828 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |