Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277486.1 | NP_001264415.1 | 6 - 7 | 531 | 124 | |
GenBank mRNA | BX933901.2 | - | 6 - 7 | 600 | 124 | |
BX934934.1 | - | 6 - 7 | 541 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |