Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015280883.2 | XP_015136369.1 | 5 - 6 | 630 | 67 | |
XM_417276.6 | XP_417276.2 | 5 - 6 | 561 | 67 | ||
GenBank mRNA | BX935679.2 | - | 5 - 6 | 547 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |