Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293389.2 | XP_015148875.1 | 4 - 5 | 1521 | 78 | |
XM_015293390.2 | XP_015148876.1 | 4 - 5 | 1520 | 78 | ||
XM_015293391.2 | XP_015148877.1 | 4 - 5 | 1521 | 78 | ||
XM_015293392.2 | XP_015148878.1 | 4 - 5 | 1531 | 78 | ||
XM_015293393.2 | XP_015148879.1 | 4 - 5 | 1519 | 78 | ||
XM_015293394.2 | XP_015148880.1 | 4 - 5 | 1524 | 78 | ||
XM_015293395.2 | XP_015148881.1 | 4 - 5 | 1654 | 78 | ||
XM_015293396.2 | XP_015148882.1 | 4 - 5 | 644 | 78 | ||
XM_025154719.1 | XP_025010487.1 | 4 - 5 | 416 | 78 | ||
GenBank mRNA | BX935948.2 | - | 2 - 3 | 179 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |