Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199565.1 | NP_001186494.1 | 1 - 2 | 94 | 78 | |
NM_001199566.1 | NP_001186495.1 | 1 - 2 | 94 | 78 | ||
XM_015286220.2 | XP_015141706.2 | 1 - 2 | 95 | 78 | ||
XM_025150663.1 | XP_025006431.1 | 1 - 2 | 485 | 78 | ||
GenBank mRNA | AM065422.1 | - | 1 - 2 | 94 | 78 | |
BX929358.2 | - | 1 - 2 | 87 | 78 | ||
BX932402.1 | - | 1 - 2 | 89 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |