Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199314.1 | NP_001186243.1 | 3 - 4 | 577 | 75 | |
XM_015283797.2 | XP_015139283.1 | 4 - 5 | 690 | 75 | ||
XM_015283838.2 | XP_015139324.1 | 4 - 5 | 728 | 75 | ||
XM_015283885.2 | XP_015139371.1 | 4 - 5 | 647 | 75 | ||
XM_015283965.1 | XP_015139451.1 | 4 - 5 | 690 | 75 | ||
XM_025147208.1 | XP_025002976.1 | 4 - 5 | 690 | 75 | ||
GenBank mRNA | BX932471.2 | - | 2 - 3 | 238 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |