Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421377.6 | XP_421377.3 | 2 | 1130 | 86 | |
GenBank mRNA | BX935378.2 | - | 1 | 755 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |