Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167728.1 | NP_001161200.1 | 3 - 4 | 580 | 84 | |
GenBank mRNA | L21928.1 | - | 3 - 4 | 453 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |