Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006345.2 | NP_001006345.1 | 2 - 3 | 145 | 71 | |
GenBank mRNA | AJ720578.1 | - | 2 - 3 | 139 | 71 | |
BU135171.1 | - | 2 - 3 | 145 | 71 | ||
CR338953.1 | - | 2 - 3 | 125 | 71 | ||
CR352384.1 | - | 1 - 2 | 115 | 71 | ||
CR353058.1 | - | 2 - 3 | 122 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |