Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006349.1 | NP_001006349.1 | 4 - 5 | 378 | 117 | |
GenBank mRNA | AJ719829.1 | - | 4 - 5 | 378 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |