Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204705.1 | NP_990036.1 | 3 - 4 | 757 | 149 | |
XM_015280241.2 | XP_015135727.1 | 3 - 4 | 800 | 149 | ||
GenBank mRNA | AF227683.1 | - | 3 - 4 | 757 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |